Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-MHC Class I (H-2Kd, H-2Dd) Mouse Monoclonal Antibody [clone: 34-1-2S]

Supplier: Bio X Cell
BE0180-100MG BE0180-50MG BE0180-1MG BE0180-5MG BE0180-25MG
103252-944EA 3920 USD InStock
103252-944 103252-946 103252-948 103252-950 103572-870 76554-368 76555-220 76555-418 76555-660 76557-372
Anti-MHC Class I (H-2Kd, H-2Dd) Mouse Monoclonal Antibody [clone: 34-1-2S]
Antibodies
InVivoMab anti-mouse MHC Class I (H-2Kd, H-2Dd), Clone: 34-1-2S, Isotype: mouse IgG2a, κ, Applications: in vivo activation of APCs

Type: Primary
Antigen: MHC Class I (H-2Kd, H-2Dd)
Clonality: Monoclonal
Clone: 34-1-2S
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty