Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Integrin beta7 Rat Monoclonal Antibody [clone: FIB504]

Supplier: Bio X Cell
BE0062-5MG BE0062-1MG BE0062-50MG BE0062-100MG BE0062-25MG
103253-582EA 3920 USD InStock
103253-582 103253-584 103253-586 103253-588 103572-498 76554-628 76555-056 76555-866 76556-250 76557-366
Anti-Integrin beta7 Rat Monoclonal Antibody [clone: FIB504]
Antibodies
InVivoMab anti-mouse/human Integrin β7, Clone: FIB504, Isotype: rat IgG2a, κ, Applications: Immunoprecipitation, Western blot, Flow cytometry

Type: Primary
Antigen: Integrin beta7
Clonality: Monoclonal
Clone: FIB504
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty