Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TIM-1 (CD365) Rat Monoclonal Antibody [clone: RMT1-10]

Supplier: Bio X Cell
BE0113-100MG BE0113-50MG BE0113-5MG BE0113-25MG BE0113-1MG
103253-724EA 3920 USD InStock
103253-724 103253-726 103253-728 103253-730 103572-532 76554-576 76555-238 76556-524 76557-022 76557-250
Anti-TIM-1 (CD365) Rat Monoclonal Antibody [clone: RMT1-10]
Antibodies
InVivoMab anti-mouse TIM-1 (CD365), Clone: RMT1-10, Isotype: rat IgG2a, κ, Applications: in vivo TIM-1 neutralization

Type: Primary
Antigen: TIM-1 (CD365)
Clonality: Monoclonal
Clone: RMT1-10
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty