Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TIM-2 Rat Monoclonal Antibody [clone: RMT2-29]

Supplier: Bio X Cell
BE0114-100MG BE0114-1MG BE0114-50MG BE0114-25MG BE0114-5MG
103253-732EA 3920 USD InStock
103253-732 103253-734 103253-736 103253-738 103572-798 76554-632 76554-916 76555-258 76555-302 76557-256
Anti-TIM-2 Rat Monoclonal Antibody [clone: RMT2-29]
Antibodies
InVivoMab anti-mouse TIM-2, Clone: RMT2-29, Isotype: rat IgM, Applications: in vivo TIM-1 neutralization

Type: Primary
Antigen: TIM-2
Clonality: Monoclonal
Clone: RMT2-29
Conjugation:
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty