Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CSF1 Rat Monoclonal Antibody [clone: 5A1]

Supplier: Bio X Cell
BE0204-1MG BE0204-100MG BE0204-50MG BE0204-5MG BE0204-25MG
103253-928EA 3920 USD InStock
103253-928 103253-930 103253-932 103253-934 103572-914 76555-062 76555-282 76555-926 76556-522 76556-968
Anti-CSF1 Rat Monoclonal Antibody [clone: 5A1]
Antibodies
InVivoMab anti-mouse CSF1, Clone: 5A1, Isotype: rat IgG1, κ, Applications: in vitro CSF1 neutralization

Type: Primary
Antigen: CSF1
Clonality: Monoclonal
Clone: 5A1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty