Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD73 Rat Monoclonal Antibody [clone: TY/23]

Supplier: Bio X Cell
BE0209-1MG BE0209-50MG BE0209-100MG BE0209-25MG BE0209-5MG
103253-952EA 3920 USD InStock
103253-952 103253-954 103253-956 103253-958 103572-920 76555-244 76555-470 76556-198 76557-114 76557-230
Anti-CD73 Rat Monoclonal Antibody [clone: TY/23]
Antibodies
InVivoMab anti-mouse CD73, Clone: TY/23, Isotype: rat IgG2a, κ, Applications: in vivo CD73 blockade

Type: Primary
Antigen: CD73
Clonality: Monoclonal
Clone: TY/23
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty