Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD209b (SIGN-R1) Armenian Hamster Monoclonal Antibody [clone: 22D1]

Supplier: Bio X Cell
BE0220-50MG BE0220-100MG BE0220-5MG BE0220-25MG BE0220-1MG
103254-024EA 3920 USD InStock
103254-024 103254-026 103254-028 103254-030 103572-568 76554-838 76555-140 76557-108 76557-290 76557-352
Anti-CD209b (SIGN-R1) Armenian Hamster Monoclonal Antibody [clone: 22D1]
Antibodies
InVivoMab anti-mouse CD209b (SIGN-R1), Clone: 22D1, Isotype: Armenian hamster IgG, κ, Applications: in vivo SIGN-R1 blockade, Immunohistochemistry (frozen), Western blot, Flow cytometry

Type: Primary
Antigen: CD209b (SIGN-R1)
Clonality: Monoclonal
Clone: 22D1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty