Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-c-myc Mouse Monoclonal Antibody [clone: 9E10]

Supplier: Bio X Cell
BE0238-1MG BE0238-25MG BE0238-50MG BE0238-100MG BE0238-5MG
103254-056EA 3920 USD InStock
103254-056 103254-058 103254-060 103254-062 103572-596 76555-012 76555-166 76555-702 76555-984 76557-232
Anti-c-myc Mouse Monoclonal Antibody [clone: 9E10]
Antibodies
InVivoMab anti-human c-myc, Clone: 9E10, Isotype: mouse IgG1, Applications: Western blot, ELISA, Flow cytometry

Type: Primary
Antigen: c-myc
Clonality: Monoclonal
Clone: 9E10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty