Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL-10R (CD210) Rat Monoclonal Antibody [clone: 1B1.3A]

Supplier: Bio X Cell
BP0050-100MG BP0050-50MG BP0050-5MG BP0050-25MG
103254-208EA 5100 USD InStock
103254-208 103254-210 103254-212 103254-214 76555-080 76555-312 76557-174 76557-184
Anti-IL-10R (CD210) Rat Monoclonal Antibody [clone: 1B1.3A]
Antibodies
InVivoPlus anti-mouse IL-10R (CD210), Clone: 1B1.3A, Isotype: rat IgG1, κ, Applications: in vivo blocking of IL-10/IL-10R signaling, in vitro blocking of IL-10R signaling, Flow cytometry

Type: Primary
Antigen: IL-10R (CD210)
Clonality: Monoclonal
Clone: 1B1.3A
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty