Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TGF-beta Mouse Monoclonal Antibody [clone: 1D11.16.8]

Supplier: Bio X Cell
BP0057-100MG BP0057-50MG BP0057-25MG BP0057-5MG
103254-216EA 5100 USD InStock
103254-216 103254-218 103254-220 103254-222 76555-570 76556-474 76556-750 76556-932
Anti-TGF-beta Mouse Monoclonal Antibody [clone: 1D11.16.8]
Antibodies
InVivoPlus anti-mouse/human/rat/monkey/hamster/canine/bovine TGF-β, Clone: 1D11.16.8, Isotype: mouse IgG1, κ, Applications: in vivo TGFβ neutralization, in vitro TGFβ neutralization

Type: Primary
Antigen: TGF-beta
Clonality: Monoclonal
Clone: 1D11.16.8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat, Monkey, Hamster, Canine, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty