Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CTLA-4 (CD152) Syrian Hamster Monoclonal Antibody [clone: 9H10]

Supplier: Bio X Cell
BP0131-50MG BP0131-25MG BP0131-100MG BP0131-5MG
103254-240EA 5305 USD InStock
103254-240 103254-242 103254-244 103254-246 76555-116 76555-970 76556-068 76556-482
Anti-CTLA-4 (CD152) Syrian Hamster Monoclonal Antibody [clone: 9H10]
Antibodies
InVivoPlus anti-mouse CTLA-4 (CD152), Clone: 9H10, Isotype: Syrian hamster IgG2, Applications: in vivo CTLA-4 neutralization, in vitro CTLA-4 neutralization

Type: Primary
Antigen: CTLA-4 (CD152)
Clonality: Monoclonal
Clone: 9H10
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: IgG2
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty