Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL-12 p40 Rat Monoclonal Antibody [clone: C17.8]

Supplier: Bio X Cell
BP0051-5MG BP0051-50MG BP0051-25MG BP0051-100MG
103254-288EA 5100 USD InStock
103254-288 103254-290 103254-292 103254-294 76554-954 76555-082 76555-288 76556-424
Anti-IL-12 p40 Rat Monoclonal Antibody [clone: C17.8]
Antibodies
InVivoPlus anti-mouse IL-12 p40, Clone: C17.8, Isotype: rat IgG2a, κ, Applications: in vivo IL-12p40 neutralization, p40 affinity chromatography, Immunoprecipitation, ELISA, Flow cytometry

Type: Primary
Antigen: IL-12 p40
Clonality: Monoclonal
Clone: C17.8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty