Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD40 Rat Monoclonal Antibody [clone: FGK4.5/FGK45]

Supplier: Bio X Cell
BP0016-2-5MG BP0016-2-50MG BP0016-2-25MG BP0016-2-100MG
103254-320EA 5100 USD InStock
103254-320 103254-322 103254-324 103254-326 76555-124 76555-396 76555-858 76557-326
Anti-CD40 Rat Monoclonal Antibody [clone: FGK4.5/FGK45]
Antibodies
InVivoPlus anti-mouse CD40, Clone: FGK4.5/FGK45, Isotype: rat IgG2a, Applications: in vivo CD40 activation, in vitro B cell stimulation/activation, in vitro DC stimulation/activation

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: FGK4.5/FGK45
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty