Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TIM-3 (CD366) Rat Monoclonal Antibody [clone: B8.2C12]

Supplier: Bio X Cell
BE0275-1MG BE0275-5MG BE0275-50MG BE0275-100MG BE0275-25MG
103254-376EA 3920 USD InStock
103254-376 103254-378 103254-380 103254-382 103572-650 76554-380 76554-712 76555-110 76556-870 76557-056
Anti-TIM-3 (CD366) Rat Monoclonal Antibody [clone: B8.2C12]
Antibodies
InVivoMab anti-mouse TIM-3 (CD366), Clone: B8.2C12, Isotype: rat IgG1, κ, Applications: in vivo TIM-3 neutralization, in vitro TIM-3 blocking, Flow cytometry

Type: Primary
Antigen: TIM-3 (CD366)
Clonality: Monoclonal
Clone: B8.2C12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty