Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-EGFR mouse Monoclonal Antibody [clone: 528 (HB-8509)]

Supplier: Bio X Cell
BE0279-50MG BE0279-5MG BE0279-1MG BE0279-25MG BE0279-100MG
103254-408EA 3920 USD InStock
103254-408 103254-410 103254-412 103254-414 103572-930 76554-422 76554-722 76555-026 76555-780 76556-948
Anti-EGFR mouse Monoclonal Antibody [clone: 528 (HB-8509)]
Antibodies
InVivoMab anti-human EGFR, Clone: 528 (HB-8509), Isotype: mouse IgG2a, Applications: in vitro EGFR blockade, Western blot, Functional assays, Immunoprecipitation, Immunohistochemistry (paraffin), Immunofluorescence, Flow cytometry

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 528 (HB-8509)
Conjugation:
Epitope:
Host: mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty