Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD40L (CD154) mouse Monoclonal Antibody [clone: 5C8]

Supplier: Bio X Cell
BE0292-1MG BE0292-50MG BE0292-100MG BE0292-25MG BE0292-5MG
103479-646EA 3920 USD InStock
103479-646 103479-648 103479-650 103479-652 103572-956 76554-330 76554-608 76555-744 76555-964 76556-844
Anti-CD40L (CD154) mouse Monoclonal Antibody [clone: 5C8]
Antibodies
InVivoMAb anti-human/monkey CD40L (CD154), Clone: 5C8, Isotype: mouse IgG2a, Applications: in vivo OX40 activation, in vitro OX40 activation, Western blot

Type: Primary
Antigen: CD40L (CD154)
Clonality: Monoclonal
Clone: 5C8
Conjugation:
Epitope:
Host: mouse
Isotype: IgG2a
Reactivity: Human, Monkey

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty