Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CSF1R (CD115) Rat Monoclonal Antibody [clone: AFS98]

Supplier: Bio X Cell
BP0213-50MG BP0213-25MG BP0213-5MG BP0213-100MG
103479-726EA 5100 USD InStock
103479-726 103479-728 103479-730 103479-732 76554-970 76556-280 76556-594 76557-304
Anti-CSF1R (CD115) Rat Monoclonal Antibody [clone: AFS98]
Antibodies
InVivoPlus anti-mouse CSF1R (CD115), Clone: AFS98, Isotype: rat IgG2a, κ, Applications: in vivo macrophage depletion, in vitro CSF-R1 neutralization, in vivo monocyte depletion, Flow cytometry

Type: Primary
Antigen: CSF1R (CD115)
Clonality: Monoclonal
Clone: AFS98
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty