Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Resveratrol

Supplier: BMT - Dr.Brandt Medizin Technik

M1606-1MG M1606-5MG
101969-314EA 294.23 USD InStock
101969-314 101969-316
Resveratrol
Resveratrol


Formula: C₁₄H₁₂O₃
MDL Number: MFCD00133799
CAS Number: 501-36-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty