Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Valganciclovir hydrochloride ≥98%, MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

M1842-1MG M1842-5MG
101969-748EA 294.23 USD InStock
101969-748 101969-750
Valganciclovir hydrochloride ≥98%, MORpure™
Valganciclovir hydrochloride


Formula: C₁₄H₂₂N₆O₅·HCl
MDL Number: MFCD08460118
CAS Number: 175865-59-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty