Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetophenone, [ring-14C(U)] ≥98%, MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MC2017-50UCI MC2017-250UCI
101970-890EA 5387.69 USD InStock
101970-890 101970-892
Acetophenone, [ring-14C(U)] ≥98%, MORpure™
Acetophenone, [ring-14C(U)]


CAS Number: 2088238-56-4
UN: 2910
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty