Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecanoic acid, [1-14C] ≥98%, MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MC2208-250UCI MC2208-50UCI MC2208-1MCI
101971-176EA 6590.77 USD InStock
101971-176 101971-174 101971-178
Dodecanoic acid, [1-14C] ≥98%, MORpure™
Dodecanoic acid, [1-14C]


CAS Number: 55322-51-5
UN: 2910
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty