Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7,10,13,16,19-Docosahexaenoic acid, [1-14C] ≥98% (by HPLC), MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MC444-10UCI MC444-50UCI MC444-250UCI
101971-900EA 1693.46 USD InStock
101971-900 101971-902 101971-904
4,7,10,13,16,19-Docosahexaenoic acid, [1-14C] ≥98% (by HPLC), MORpure™
4,7,10,13,16,19-Docosahexaenoic acid, [1-14C]


CAS Number: 119798-49-1
UN: 2910
ADR: ,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty