Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Saquinavir, [3H]

Supplier: BMT - Dr.Brandt Medizin Technik

MT1731-50UCI MT1731-250UCI
101972-508EA 2595.77 USD InStock
101972-508 101972-510
Saquinavir, [3H]
Saquinavir, [3H]


CAS Number: 127779-20-8
UN: 2910
ADR: ,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty