Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluoroacetic acid, potassium salt, [3H] ≥97%, MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MT1967-50UCI MT1967-250UCI
101972-894EA 1601.92 USD InStock
101972-894 101972-896
Fluoroacetic acid, potassium salt, [3H] ≥97%, MORpure™
Fluoroacetic acid, potassium salt, [3H]


CAS Number: 23745-86-0
UN: 2910
ADR: ,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty