Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Deoxy-3'-fluorothymidine, [methyl-3H] ≥97% (by HPLC), MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MT645-250UCI MT645-1MCI
101973-286EA 2700.38 USD InStock
101973-286 101973-288
3'-Deoxy-3'-fluorothymidine, [methyl-3H] ≥97% (by HPLC), MORpure™
3'-Deoxy-3'-fluorothymidine, [methyl-3H]


CAS Number: 2086327-39-9
UN: 2910
ADR: ,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty