Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Suramin sodium, [3H] ≥97% (by HPLC), MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MT823-100UCI MT823-250UCI MT823-1MCI
101973-694EA 2026.92 USD InStock
101973-694 101973-696 101973-698
Suramin sodium, [3H] ≥97% (by HPLC), MORpure™
Suramin sodium, [3H]


CAS Number: 339532-33-1
UN: 2910
ADR: ,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty