Human Recombinant NRG1-B (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC.
100-46AF-10UG 100-46AF-50UG 100-46AF-100UG 100-46AF-500UG 100-46AF-1MG
76046-792EA 90.25 USD InStock
76046-792 76046-794 76046-796 76046-798 76046-800
Human Recombinant NRG1-B (Animal free) (from E. coli)
Proteins and Peptides
Visfatin is an adipokine produced by visceral adipose tissue.  Visfatin acts as a pro-inflammatory factor for peripheral blood mononuclear cells (PBMCs) and mimics the effects of insulin on adipocytes, monocytes, and hepatocytes.
  • High quality
  • Low endotoxin
  • Affordable
Neuregulin 1-beta (NRG1-β) is one of many alternatively-spliced isoforms of the NRG1 gene and contains a soluble EGF-like domain. The EGF-like domain of NRG1-β signals through the ErbB2, ErbB3, and ErbB4 receptor tyrosine kinases. NRG1-β is an important growth factor involved in neuroinflammation, nerve regeneration, and cardiovascular processes. 
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty