Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-L-phenylalanine methyl ester hydrochloride ≥99% (by HPLC)

Supplier: A.W. SCHÜRR GmbH

14288-1G 14288-5G 14288-25G
102594-328EA 109.34 USD InStock
102594-328 102594-330 102637-106
2-Fluoro-L-phenylalanine methyl ester hydrochloride ≥99% (by HPLC)
2-Fluoro-L-phenylalanine methyl ester hydrochloride
Synonyms: L-Phe(2-F)-OMe·HCl, o-Fluoro-L-Phe-OMe·HCl, (S)-2-amino-3-(2-fluorophenyl) propionic acid.


Formula: C₁₀H₁₃ClFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08458514
CAS Number: 457654-69-2
Order Now
Specification Test Results
Melting Point 177 - 182 ºC
Purity ≥ 99% (HPLC)
Optical Rotation [a]D25 = + 7 ± 2 º (C=1 in H2O)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty