BRIGHTSORB™ High Visibility Spill Kit, Brady Worldwide
Supplier: Brady Worldwide
148627 148625
76046-574KT 491.68 USD InStock
76046-574 76046-576 76046-578 76046-580 76046-582
BRIGHTSORB™ High Visibility Spill Kit, Brady Worldwide
Spill Control Materials
Spill Kits combine absorbents, accessories, PPE, and instructional guidelines to ensure preparation for emergencies when a spill occurs.
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Slip, trip, and fall hazard warning printed right on the absorbent alerts employees to potential danger
  • Improve worker productivity and efficiency by placing spill kits around any facility, close to areas prone to spills
The 5 Gallon Kit includes (10) 15" x 19" pads; (2) 3" x 4' socks; (1) disposal bag; Nitrile gloves; Instruction sheet.
The 6.5 Gallon Kit includes (10) 15" x 19" pads; (3) 3" x 4' socks; (2) disposal bags; Nitrile gloves; Instruction sheet.
The 20 Gallon Kit includes (12) 15" x 19" pads; (3) 3" x 12' socks; (2) 18" x 18" pillows; (3) disposal bags; Nitrile Gloves; Goggles; Emergency Response Guidebook.
The 55 Gallon Kit includes (50) 15" x 19" pads; (4) 3" x 12' socks; (8) 18" x 18" pillows; (5) disposal bags; Goggles; Nitrile Gloves; Emergency Response Guidebook.
The 95 Gallon Kit includes (110) 15" x 19" pads; (12) 3" x 4' socks; (8) 3" x 12' socks; (8) 18" x 18" pillows; (10) disposal bags; Nitrile gloves; Goggles; Emergency Response Guidebook.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty