Mitis Salivarius Agar Base, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M259-2.5KG M259-5KG
89224-954EA 990.26 USD InStock
89224-954 89224-956
Mitis Salivarius Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For isolation of streptococci from mixed cultures, especially Streptococcus Mitis, Streptococcus Salivarius and Streptococcus Faecalis.
  • Multiple sizes available
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty