BD BBL™ Trypticase™ Soy Agar Settling Plates, BD Diagnostics
Supplier: Seca Med.Waagen u.Meßsysteme
292398 222205 222206
90001-272PK 39.02 USD InStock
90001-272 90004-282 90001-270 90001-610 90004-174 90001-612
BD BBL™ Trypticase™ Soy Agar Settling Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD BBL™ prepared agar plates in sterile packs are plated media in gamma-irradiated packs that are validated to performance and sterility according to AAMI (Association for the Advancement of Medical Instrumentation) guidelines.
  • For critical environments
  • Validated to 10⁻⁶ SAL
  • Sterile inner wrapping
Outer Tyvek/polyethylene wrapping minimizes the risk of false contamination, provides fiber-free bacterial barrier and allows for gas exchange. To use, remove the outer wrapping in the non-critical environment, then remove the sterile inner wrapping in the clean area; the included bag may be used to transport plates from your critical environment. Storage temperature: 2 to 8 °C.
Settling plates for active and passive air sampling.
Información para pedidos: Each pack or case contains a signed, lot-specific certificate of analysis.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty