Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

JEFFAMINE® ED-2003 amine 50% (v/v)

Supplier: QGT Quarzglastechnik GmbH & Co KG

1008030 1008031
101443-308EA 108.54 USD InStock
101443-308 101443-656
JEFFAMINE® ED-2003 amine 50% (v/v)
JEFFAMINE® ED-2003 amine


Formula: CH₃CH(NH₂)CH₂[OCH(CH₃)CH₂]l(OCH₂CH₂)m[OCH₂CH(CH₃)]nNH₂
MW: 2000 g/mol
MDL Number: MFCD00147794
CAS Number: 65605-36-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty