Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium fluoride 1 M

Supplier: QGT Quarzglastechnik GmbH & Co KG

1008199 1008198
101443-670EA 162.15 USD InStock
101443-670 101443-322
Potassium fluoride 1 M
Potassium fluoride
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.


Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505
Melting Pt: 860
MDL Number: MFCD00011398
CAS Number: 7789-23-3
UN: 1812
ADR: 6.1 III
Merck Index: 13,07714
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty