Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyethylene glycol 1,000 50% (w/v)

Supplier: QGT Quarzglastechnik GmbH & Co KG

1008052 1008053
101443-450EA 99.38 USD InStock
101443-450 101443-840
Polyethylene glycol 1,000 50% (w/v)
Polyethylene glycol 1,000
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.


Formula: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Flash Pt: 260
MDL Number: MFCD00081839
CAS Number: 25322-68-3
Merck Index: 13,07651
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty