Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium dihydrogen phosphate, anhydrous 99.5-100.5%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO buffer substance, Fluka™

Supplier: BURDICK & JACKSON FKA SIGMA

60220-1KG 60220-2.5KG 60220-500G 60220-5KG
BJ60220-1KGEA 229 USD InStock
BJ60220-1KG BJ60220-2.5KG BJ60220-500G BJ60220-5KG
Potassium dihydrogen phosphate, anhydrous 99.5-100.5%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO buffer substance, Fluka™
Potassium dihydrogen phosphate


Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: 100
Melting Pt: 253 °C
Density: 2.34 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
Merck Index: 13,07744
Order Now
Specification Test Results
Assay (alk., calculated to dried substances) 99.5 - 100.5 %
Assay Min. 99.0 %
Loss on drying (130°C) Max. 0.2 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 4.2 - 4.5
Arsenic (As) Max. 0.5 ppm
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0015 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 5 ppm
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Sodium (Na) Max. 50 ppm
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm
Total N Max. 0.001 %
KMnO₄ reducing matter (as O) complying
Appearance of the solution complying
Appearance (color) complying
Appearance (form) complying

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty