Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Human Beta-Amyloid (2-42)

Supplier: Hytest Ltd

AS-29909-01 AS-29909-1
103000-894EA 182.69 USD InStock
103000-894 103000-896
Human Beta-Amyloid (2-42)
Proteins and Peptides
This peptide is beta-amyloid (1-42) N-terminally truncated. It was shown that supplementing the media with N-terminally truncated Abeta (2-40) and (2-42) induce the phagocytosis of polystyrene particles by primary human monocytes. N-terminally truncated Aβ(x–42) induced the phagocytosis of PSPs significantly more effectively than did Aβ(x–40).
Sequence: AEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
Molecular Weight: 4399 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty