Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Human Beta-Amyloid (1-40)

Supplier: Hytest Ltd

AS-24235- AS-24236 AS-24236-5
102999-790EA 247 USD InStock
102999-790 102999-792 102999-794
Human Beta-Amyloid (1-40)
Proteins and Peptides
Aß (1-40) together with Aß (1-42) are two major C-terminal variants of the Aß protein constituting the majority of Aßs. These undergo post-secretory aggregation and deposition in the Alzheimer’s disease brain.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV
Molecular Weight: 4329.9 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty