Human LC-Beta-Amyloid (1-40),Biotin
Supplier: Hytest Ltd
AS-24645 AS-24645-01 AS-24648
102999-820EA 1107.85 USD InStock
102999-820 103003-796 103003-798
Human LC-Beta-Amyloid (1-40),Biotin
Proteins and Peptides
This biotinylated Aß(1-40) contains a 6-carbon long chain (LC) to provide more accesibility for avidin attachment.
  • Peptide Content: ≥ 60%
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty