Pierce™ NeutrAvidin™ Protein, Thermo Scientific
Supplier: Paul Schöndorf Metallwaren GmbH
31000 31001 31002 31006 31007
PI31000EA 285.49 USD InStock
PI31000 PI31001 PI31002 PI31006 PI31007
Pierce™ NeutrAvidin™ Protein, Thermo Scientific
Protein Purification Protein Purification Reagents
Pierce™ NeutrAvidin™ Protein is a specially-prepared form of avidin biotin-binding protein that decreases background in biotin-binding.
  • Near-neutral isoelectric point - pI = 6.3
  • Nearly devoid of glycosylation - decreased possibility of lectin binding compared to native avidin
  • No RYD recognition sequence - no known off-target binding domains like streptavidin
  • Less expensive than streptavidin
NeutrAvidin Protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. Purified NeutrAvidin Protein provides exceptional performance in western blot, ELISA, and IHC applications that require biotin-binding probes. Assay specificity, sensitivity, and signal-to-noise ratios with this protein are generally equivalent or better than with the significantly more expensive streptavidin. The specific activity for biotin-binding is approximately 14 µg/mg of protein, which is near the theoretical maximum activity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty