Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[Arg8]-Vasopressin (AVP) Peptide

Supplier: Hytest Ltd

AS-24289 AS-24290
102996-516EA 77.46 USD InStock
102996-516 102996-518
[Arg8]-Vasopressin (AVP) Peptide
Proteins and Peptides
AVP have been identified as an important regulator of fluid and electrolyte homeostasis through its anti-diuretic action on the kidney, After myocardial infarction, plasma levels of [Arg8]-vasopressin rise to recover hemodynamics.
Sequence: CYFQNCPRG-NH2 (Disulfide bridge: 1-6)
MW: 1084.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C


Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty