Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: BURDICK & JACKSON FKA SIGMA
Danger

33211-180KG 33211-25L 33211-1L 33211-2.5L 33211-4X2.5L 33211-6X1L
BJ33211-180KGEA 5512.17 USD InStock
BJ33211-180KG BJ33211-25L BJ33211-35KG BJ33211-1L BJ33211-2.5L BJ33211-4X2.5L BJ33211-6X1L
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Ethyl acetate
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602


Formula: CH₃COOC₂H₅
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: –84 °C
Density: 0.901 g/cm³
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=0f6c525db7a2f3725affe5045040d70e0ac99352&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) Min. 99.5 %
Boiling range 76 - 78 °C
Density (D 20/4) 0.899 - 0.901
Refractive index (n 20/D) 1.3710 - 1.3730
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.0045 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Molybdenum (Mo) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
methyl acetate Max. 0.1 %
Ethanol (GC) Max. 0.1 %
Isobutyl alcohol (GC) Max. 0.1 %
Methanol (GC) Max. 0.1 %
Reaction against H₂SO₄ complying
APHA Max. 10

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty