High Pressure Analytical Static Mixers, Perkinelmer
Supplier: PERKINELMER U.S. LLC
N2911200 N2911201 N2911202 N2911203 N2911205
101692-328EA 1000.45 USD InStock
101692-328 101691-248 101692-326 97019-224 97019-226 97019-228 97019-230 97019-232
High Pressure Analytical Static Mixers, Perkinelmer
Shakers and Mixers Vortex Mixers
Static Mixers are important in facilitating complete mobile phase blending, resulting in improved retention performance. These mixers should be used as part of FX10, or FX15 LC systems, where high pressure blending is required.
Mixers incorporate a highly efficient cross flow shearing mechanism which produces vortex sheer mixing over a wide range of volumes. They are now offered in 18000 psi pressure max. flavor.
A good first choice selection is a volume of about half the flow rate volume. For example, when pumping at a 0.5 ml/min flow rate, a 350 μl mixer provides good mixing with very good baseline stability.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty