Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ferritin (equine from horse spleen, thick), dark brown liquid

Supplier: E. Becker & Co. GmbH

0215111980 0215111990
76102-706EA 128.58 USD InStock
76102-706 76102-708
Ferritin (equine from horse spleen, thick), dark brown liquid
Ferritin
Ferritin, Equine is a shell of 24 protein subunits (apoferritin) and a core Fe 3+ ions with a peptide MW of 440 kDa. Review of the ferritin-like superfamily of iron-storage proteins, virtually ubiquitous in life on earth.
A ubiquitous iron storage protein that plays a key role in iron metabolism. It serves as an intracellular iron reserve (particularly in spleen, liver, intestinal mucosa, and bone marrow) and functions in iron detoxification. Studies have shown that ferritin iron incorporation is mediated by a ferroxidase activity associated with ferritin H subunits and a nucleation center associated with ferritin L subunits. Release of iron from ferritin has an essential role in iron-dependent lipid peroxidation and may contribute to free radical-induced cell damage in vivo. Therefore, by binding iron, ferritin may function as an antioxidant.
Store at +4 °C.

  • Presentation: Thick, Dark Brown Liquid
  • Formulation: In 150 mM NaCl, 0.002% sodium azide
  • 2X Crystallized, Essentially cadmium-free, but may contain up to 1% (as % of ferritin)
  • Soluble in 0.15 M sodium chloride, distilled water or dilute buffer


Formula: C₁₆H₃₃O₂S⁻
MW: 289.498 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00081593
CAS Number: 9007-73-2
Order Now
Specification Test Results
Identity Test Passes
Source Horse Spleen

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty