Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Pseudouridine (synthetic)

Supplier: E. Becker & Co. GmbH

0219917810 0219917850 0219917883
76177-492EA 303.23 USD InStock
76177-492 76177-494 76177-496
β-Pseudouridine (synthetic)
β-Pseudouridine
β-Pseudouridine isomer of the nucleoside uridine found in all species and in many classes of RNA except mRNA. It is formed by enzymes called Ψ synthases, which post-transcriptionally isomerize specific uridine residues in RNA in a process termed pseudouridylation. Studies suggest that β-Pseudouridine reduces radiation-induced chromosome aberrations in human lymphocytes.
+4 °C

  • Presentation: Off-White Powder
  • Soluble in water (50 mg/mL - clear, colorless solution).


Formula: C₉H₁₂N₂O₆
Boiling Pt: 598.4 °C (760 mmHg)) (lit.)
Density: 1.64 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00038458
CAS Number: 1445-07-4
Order Now
Specification Test Results
Identity Test Passes
Source Synthetic

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty