Ugo Basile Rota-Rod, Stoelting
Supplier: Stoelting
57626 57624 57625
10842-392EA 19429.28 USD InStock
10842-392 10842-406 10842-408
Ugo Basile Rota-Rod, Stoelting
Animal Supplies Animal Environmental Enrichment
Ugo Basile Rota-Rod has a great value in research involving screening of drugs which are potentially active on motory coordination.
  • Excellent in assessing the effects of drugs, brain damage, and diseases on motor coordination and fatigue
  • Does not make noise
  • The angular speed can be preset from 2 – 80 rpm in steps of 1 rpm
  • In the acceleration mode, the speed changes can be preset in 6 second (10 minute intervals in 6 second steps)
  • The tiltable digital panel shows accrued rotations and time on the rod
When the animal falls off its cylinder section onto the plate below, the plate trips, thereby recording the animal’s endurance time in seconds. The reverse rotation can be selected, which takes place at minimum speed, at the end of a programmable acceleration-deceleration sequence. A rocking motion is also presettable, with adjustable angular amplitude, speed, and acceleration. The Rota-Rod includes software for connection to PC, and parameters can be controlled via your PC.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty