Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(-)-Sorbitol 91.0-100.5% (dry basis), Ultrapure

Supplier: THERMO SCIENTIFIC CHEMICALS INC MS

J21710-A7 J21710-36
101172-996EA 168.23 USD InStock
101172-996 101173-910 101175-736
D-(-)-Sorbitol 91.0-100.5% (dry basis), Ultrapure
D(-)-Sorbitol


Formula: HOH₂C(CH(OH))₄CH₂OH
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 14,8725
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (Dry Basis) 91.0 to 100.5%
Assay Reducing Sugars ≤0.3%
Impurity Content Limit of Nickel ≤1 ppm
Impurity Content Total Molds and Yeasts ≤100 cfu/g
Water Content (Karl Fischer Titration) ≤1.5%
pH (10%, H₂O) 3.5 - 7.0
Identification (FTIR) Conforms
Identity (A, B) Passes Tests
Total Aerobic Plate Count ≤1000 cfu/g
Residue on Ignition ≤0.1%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty