Human Recombinant Relm- Alpha
PRT220-0025 PRT220-0100
76202-244EA 422 USD InStock
76202-244 76288-386
Human Recombinant Relm- Alpha
Proteins and Peptides
Produced in E. coli. A non-glycosylated protein containing 88 amino acids.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty