Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BriskHeat® Silicone-Rubber Heater Blankets with Control and Adhesive, for Metal Surfaces, 120VAC, Cole-Parmer

Supplier: Argos Technologies
36001-98 36002-01 36002-05 36002-09 36002-13 36002-17 36002-21 36002-25 36002-29
76181-604EA 314.12 USD InStock
76181-604 76181-606 76181-608 76181-610 76181-612 76181-614 76181-616 76181-618 76181-620
BriskHeat® Silicone-Rubber Heater Blankets with Control and Adhesive, for Metal Surfaces, 120VAC, Cole-Parmer
Heaters Heating Mantles
Rugged, moisture/chemical resistant heaters provide uniform heat distribution across large surfaces.

  • Maximum exposure temperature of 425 °F (218 °C)
  • Minimum temperature of 50 °F (10 °C)
  • Lead length 72"
  • US Standard plug type
  • Easy-to-apply industrial strength pressure sensitive adhesive backing

These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks -More Product Information
Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty