Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Biotin Mouse Monoclonal Antibody (CF647) [clone: Hyb8]

Supplier: Biotium
BNC470400-100 BNC470400-500
75885-350EA 205.77 USD InStock
75885-350 75885-352
Anti-Biotin Mouse Monoclonal Antibody (CF647) [clone: Hyb8]
Antibodies
This antibody recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone Hyb8
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty