Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ITGB4 Mouse Monoclonal Antibody (CF488A) [clone: UM-A9]

Supplier: Biotium
BNC880449-100 BNC880449-500
75883-380EA 205.77 USD InStock
75883-380 75883-382
Anti-ITGB4 Mouse Monoclonal Antibody (CF488A) [clone: UM-A9]
Antibodies
This antibody recognizes a protein of 205 kDa, which is identified as integrin beta-4 (ITGB4). Its epitope is localized in the extracellular domain of ITGB4 protein. Integrins are heterodimers comprised of alpha and beta subunits, that are non-covalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta-4 subunit, a receptor for the laminins. This subunit tends to associate with alpha-6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: Clone UM-A9
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty